Anti-CALR Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-068EA 480 EUR
PRSI49-068
Anti-CALR Mouse Monoclonal Antibody
Antikörper
Calreticulin antibody can be used in ELISA starting at 1:2000 - 1:10000, Western Blot starting at 1:500 - 1:3000, immunohistochemistry starting at 5 μg/mL, immunofluorescence, and immunoprecipitation.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CALR
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Gibbon, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR