Anti-FER Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-062EA 480 EUR
PRSI49-062
Anti-FER Mouse Monoclonal Antibody
Antikörper
FER antibody can be used in ELISA starting at 1:15000 - 1:75000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 10 μg/mL, and immunofluorescence.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FER
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR