Anti-S100 B Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-060EA 375 EUR
PRSI49-060
Anti-S100 B Mouse Monoclonal Antibody
Antikörper
S100B antibody can be used in ELISA, Western Blot starting at 1:200, and immunohistochemistry starting at 5 μg/mL.

0.03% sodium azide.

0.03% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: S100 B
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR