Anti-CK5 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-058EA 480 EUR
PRSI49-058
Anti-CK5 Mouse Monoclonal Antibody
Antikörper
Cytoke antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 2.5 μg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CK5
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR