Anti-FGFR4 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-057EA 480 EUR
PRSI49-057
Anti-FGFR4 Mouse Monoclonal Antibody
Antikörper
FGFR4 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 2.5 μg/mL.

0.03% sodium azide.

0.03% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FGFR4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR