Anti-ICAM1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-050EA 619 EUR
PRSI49-050
Anti-ICAM1 Mouse Monoclonal Antibody
Antikörper
ICAM1 antibody can be used in Western Blot starting at 0.5 - 4 μg/mL, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, pH 7.4, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ICAM1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR