Anti-ACTB Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-048EA 375 EUR
PRSI49-048
Anti-ACTB Mouse Monoclonal Antibody
Antikörper
Beta-actin antibody can be used in Western Blot, and immunohistochemistry starting at 10 μg/mL.

0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ACTB
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR