Anti-MUC2 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-047EA 480 EUR
PRSI49-047
Anti-MUC2 Mouse Monoclonal Antibody
Antikörper
MUC2 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

10mM PBS/1% BSA buffer pH 7.4 with 0.1% sodium azide.

Store MUC2 antibody at 2 °C to 8 °C degrees. Do not freeze.

Type:
Antigen: MUC2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR