Anti-SOCS3 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-045EA 514 EUR
PRSI49-045
Anti-SOCS3 Mouse Monoclonal Antibody
SOCS3 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 2.5 μg/mL, and immunoprecipitation

Protein A Column

PBS, pH 7.4, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SOCS3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR