Anti-CD34 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-041EA 480 EUR
PRSI49-041
Anti-CD34 Mouse Monoclonal Antibody
CD34 antibody can be used in immunohistochemistry starting at 10 μg/mL, and flow cytometry starting at 1:25 - 1:100.

Purified

PBS, 0.1% sodium azide and a stabilizing agent.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CD34
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR