Anti-INS Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-040EA 480 EUR
PRSI49-040
Anti-INS Mouse Monoclonal Antibody
Antikörper
Insulin antibody can be used in Western Blot, and immunohistochemistry starting at 10 μg/mL.

Purified

PBS, 0.1% sodium azide, pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: INS
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR