Anti-ZAP70 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-036EA 480 EUR
PRSI49-036
Anti-ZAP70 Mouse Monoclonal Antibody
Antikörper
ZAP70 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, pH 7.2, 0.09% azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ZAP70
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR