Anti-CD44 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-031EA 0 EUR
PRSI49-031
Anti-CD44 Mouse Monoclonal Antibody
CD44 antibody can be used in Western Blot, and immunohistochemistry starting at 20 μg/mL.

Protein A Column

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CD44
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR