Anti-BSG Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-026EA 480 EUR
PRSI49-026
Anti-BSG Mouse Monoclonal Antibody
CD147 antibody can be used in Western Blot, and immunohistochemistry starting at 5 μg/mL.

Protein A Column

Azide free PBS, preserved by filter sterilization, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BSG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR