Anti-CK19 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-020EA 480 EUR
PRSI49-020
Anti-CK19 Mouse Monoclonal Antibody
Cytoke antibody can be used in Western Blot starting at 1:1000, and immunohistochemistry starting at 1:50.

Protein A Column

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CK19
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR