Anti-LAMP2 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-019EA 480 EUR
PRSI49-019
Anti-LAMP2 Mouse Monoclonal Antibody
Antikörper
LAMP2 antibody can be used in Western Blot at 1:1000, immunohistochemistry starting at 1:50, and immunoprecipitation starting at 1:50.

Protein G Column

PBS, pH 7.4, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LAMP2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR