Anti-THY1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-018EA 480 EUR
PRSI49-018
Anti-THY1 Mouse Monoclonal Antibody
THY1 antibody can be used in ELISA, immunohistochemistry starting at 1:100, immunocytochemistry, immunofluorescence, and flow cytometry.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide.

Aliquot and store at 4ºC. Avoid freezing.

Type:
Antigen: THY1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Baboon, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR