Anti-BSG Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-017EA 480 EUR
PRSI49-017
Anti-BSG Mouse Monoclonal Antibody
CD147 antibody can be used in ELISA starting at 1:2000 - 1:12000, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 1:500 - 1:2000.

Protein G Column

PBS, pH 7.4, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BSG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR