Anti-SERPINA1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-015EA 375 EUR
PRSI49-015
Anti-SERPINA1 Mouse Monoclonal Antibody
Antikörper
SERPINA1 antibody can be used in Western Blot, and immunohistochemistry starting at 10 μg/mL.

Protein A Column

PBS, pH 7.4, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SERPINA1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR