Anti-F8 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-014EA 480 EUR
PRSI49-014
Anti-F8 Mouse Monoclonal Antibody
Antikörper
Factor VIII antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, pH 7.4

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: F8
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR