Anti-MAT2A Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-008EA 619 EUR
PRSI49-008
Anti-MAT2A Mouse Monoclonal Antibody
Antikörper
MAT2A antibody can be used in Western Blot starting at 5 - 2 μg/mL, and immunohistochemistry starting at 20 μg/mL.

Protein G Column

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MAT2A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR