Anti-RBP4 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-006EA 480 EUR
PRSI49-006
Anti-RBP4 Mouse Monoclonal Antibody
Antikörper
RBP4 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 1 - 5 μg/mL.

Purified

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RBP4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR