Anti-CD99 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-999EA 480 EUR
PRSI48-999
Anti-CD99 Mouse Monoclonal Antibody
CD99 antibody can be used in ELISA starting at 2 - 4 μg/mL, immunohistochemistry starting at 5 μg/mL, and neutralization.

Precipitation

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at 4ºC. Avoid freezing.

Type:
Antigen: CD99
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR