Anti-CD59 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-992EA 375 EUR
PRSI48-992
Anti-CD59 Mouse Monoclonal Antibody
Antikörper
CD59 antibody can be used in immunohistochemistry starting at 15 μg/mL.

Protein A Column

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CD59
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR