Anti-CD7 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-987EA 480 EUR
PRSI48-987
Anti-CD7 Mouse Monoclonal Antibody
Antikörper
CD7 antibody can be used in immunohistochemistry starting at 20 μg/mL.

Ion Exchange Chromatography

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CD7
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR