Anti-CK8 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-985EA 375 EUR
PRSI48-985
Anti-CK8 Mouse Monoclonal Antibody
Antikörper
KRT8 antibody can be used in ELISA, and immunohistochemistry starting at 5 μg/mL.

Protein A Column

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CK8
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR