Anti-ATG5 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-982EA 480 EUR
PRSI48-982
Anti-ATG5 Rabbit Polyclonal Antibody
Antikörper
ATG5 is required for the process of autophagy, an intracellular process for bulk degradation of cytoplasmic components. ATG5 conjugates to ATG12 and associates with isolation membranes to form autophagosomes. The autophagosomes then enclose cytoplasmic components and deliver them to lysosomes/vacuoles to be degraded. ATG5 has been shown to be involved in apoptosis with expression late in the apoptotic process downstream of caspase activity.

ATG5 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 μg/mL.

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ATG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Pufferfish, Pig, Rat, Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR