Anti-RAMP3 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-972EA 480 EUR
PRSI48-972
Anti-RAMP3 Rabbit Polyclonal Antibody
Antikörper
RAMP3 is a member of the RAMP family of single-transmembrane-domain proteins, called receptor-activity-modifying proteins. RAMPs are type I transmembrane proteins with an extracellular N terminus and a cytoplasmic C terminus. RAMPs are required to transport calcitonin-receptor-like receptor (CRLR) to the plasma membrane. CRLR, a receptor with seven transmembrane domains, can function as either a calcitonin-gene-related peptide (CGRP) receptor or an adrenomedullin receptor, depending on which members of the RAMP family are expressed. Co-expression of RAMP3 with CRLR results in the production of a functional adrenomedullin receptor.

RAMP3 antibody can be used in ELISA starting at 1:3000 - 1:6000, Western Blot starting at 1:3000 - 1:6000, immunohistochemistry starting at 10 μg/mL, and immunoprecipitation

PBS, 0.01% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RAMP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR