Anti-NOV Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-970EA 480 EUR
PRSI48-970
Anti-NOV Rabbit Polyclonal Antibody
Antikörper
NOV antibody can be used in Western Blot starting at 2 μg/mL, and immunohistochemistry starting at 10 μg/mL.

PBS, 0.01% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NOV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR