Anti-MAPK9 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-968EA 480 EUR
PRSI48-968
Anti-MAPK9 Rabbit Polyclonal Antibody
Antikörper
JNK2, a MAPK type protein kinase, regulates cell apoptosis, differentiation and growth in response to growth factors, inflammatory cytokines and cellular stress. JNK2 is phosphorylated by by MKK4/MAP2K4 and MKK7/MAP2K7. Activated JNK2 translocates to the nucleus where it phosphorylates transcription factors, including c-Jun, ATF2, ELK1, Jun-D and NFAT4. At least four distinct isoforms, resulting from alternatively spliced transcript variants, have been reported - three different isoforms comprised of 382 amino acids and one comprised of 424 amino acids.

MAPK9 antibody can be used in Western Blot starting at 2 μg/mL, immunohistochemistry starting at 5 μg/mL, immunocytochemistry, and immunoprecipitation

PBS, 0.01% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MAPK9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR