Anti-ACSL3 Rabbit Polyclonal Antibody
Lieferant: ProSci
PRSI48-959EA 480 EUR
PRSI48-959
Anti-ACSL3 Rabbit Polyclonal Antibody
ACSL3 antibody can be used in Western Blot starting at 1 - 3 μg/mL, and immunohistochemistry starting at 10 μg/mL.
PBS, 0.01% sodium azide, 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ACSL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human