Anti-CXCR7 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-954EA 619 EUR
PRSI48-954
Anti-CXCR7 Rabbit Polyclonal Antibody
Antikörper
RDC1 was originally considered to be an Adrenomedullin receptor that binds calcitonin gene-related peptide (CGRP) and adrenomedullin. RDC1 and GPRN1 represent two tissue-specific subtypes of the same 60- kDa receptor. RDC1 has the structural characteristics of a chemokine receptor. Recently it has been shown that RDC1 binds to the chemokine CXCL12 and should be considered to be a chemokine receptor.

CXCR7 antibody can be used in immunohistochemistry, Western Blot starting at 1:500, and immunohistochemistry starting at 1:100.

Protein G Column

PBS, 0.05% BSA, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CXCR7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR