Anti-GRK5 Rabbit Polyclonal Antibody
Lieferant: ProSci
PRSI48-952EA 480 EUR
PRSI48-952
Anti-GRK5 Rabbit Polyclonal Antibody
GRK5 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.
PBS, 0.2% gelatin, 0.05% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GRK5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Human, Mouse, Pufferfish, Rat, Zebrafish