Anti-MAF Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-948EA 480 EUR
PRSI48-948
Anti-MAF Rabbit Polyclonal Antibody
MAF antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MAF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Bovine, Dog, Gorilla, Human, Monkey, Mouse, Rat, Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR