Anti-HSP90AB1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-947EA 480 EUR
PRSI48-947
Anti-HSP90AB1 Rabbit Polyclonal Antibody
Antikörper
HSP90B1 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HSP90AB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chimpanzee, Chicken, Xenopus, Horse, Human, Monkey, Mouse, Rat, Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR