Anti-HIPK3 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-944EA 480 EUR
PRSI48-944
Anti-HIPK3 Rabbit Polyclonal Antibody
Antikörper
HIPK3, a nuclear serine/threonine kinase similar to the MNB/DYRK type protein kinases. HIPK3 may be involved in growth control pathways and in multidrug resistance. HIPK3 interacts with Fas and FADD and modulates FasL-induced JNK activation.

HimmunoprecipitationK3 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HIPK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR