Anti-PPP1R1A Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-938EA 480 EUR
PRSI48-938
Anti-PPP1R1A Rabbit Polyclonal Antibody
Antikörper
PPP1R1A antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 10 - 20 μg/mL, and immunofluorescence starting at 10 μg/mL.

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PPP1R1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chimpanzee, Dog, Xenopus, Horse, Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR