Anti-GPBAR1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-937EA 0 EUR
PRSI48-937
Anti-GPBAR1 Rabbit Polyclonal Antibody
BG37/TGR5 is the first Bile Acid GPCR to be identified.

GPBAR1 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPBAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR