Anti-HCAR3 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-921EA 480 EUR
PRSI48-921
Anti-HCAR3 Rabbit Polyclonal Antibody
GPR109B/HM74 is a Nicotinic Acid GPCR.

GPR109B antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HCAR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Dog, Hamster, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR