Anti-FNTA Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-919EA 480 EUR
PRSI48-919
Anti-FNTA Rabbit Polyclonal Antibody
FNTA antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 2.5 μg/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FNTA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chimpanzee, Chicken, Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR