Anti-GPR77 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-918EA 480 EUR
PRSI48-918
Anti-GPR77 Rabbit Polyclonal Antibody
Antikörper
C5L2 is an Anaphylatoxin Receptor. Binding to anaphylatoxins can result in increased triglyceride synthesis, although its precise role in cellular responses has not been determined.

GPR77 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 10 μg/mL.

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPR77
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chimpanzee, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR