Anti-KDM1A Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-917EA 480 EUR
PRSI48-917
Anti-KDM1A Rabbit Polyclonal Antibody
AOF2 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: KDM1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR