Anti-TMBIM6 Mouse Monoclonal Antibody
Lieferant: ProSci
PRSI48-914EA 514 EUR
PRSI48-914
Anti-TMBIM6 Mouse Monoclonal Antibody
TEGT antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 μg/mL.
Protein G Column
PBS, 0.2% gelatin, 0.05% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TMBIM6
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat