Anti-NPY2R Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-910EA 480 EUR
PRSI48-910
Anti-NPY2R Rabbit Polyclonal Antibody
Antikörper
NPY2R belongs to the Neuropeptide Receptor GPCR family.

NPY2R antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NPY2R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Monkey, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR