Anti-RIPK3 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-909EA 514 EUR
PRSI48-909
Anti-RIPK3 Rabbit Polyclonal Antibody
RIPK3 belongs to the RIP subfamily of kinases. RIP3 binds RIP through this unique C-terminal segment to inhibit RIP and TNFR1 mediated NF-κB activation and induce apoptosis.

RimmunoprecipitationK3 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 10 μg/mL.

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RIPK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR