Anti-MCTS1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-875EA 514 EUR
PRSI48-875
Anti-MCTS1 Rabbit Polyclonal Antibody
MCTS1 antibody can be used in ELISA, and immunohistochemistry starting at 10 μg/mL.

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MCTS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gibbon, Bovine, Chicken, Dog, Xenopus, Human, Mouse, Pig, Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR