Anti-DCBLD2 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-871EA 480 EUR
PRSI48-871
Anti-DCBLD2 Rabbit Polyclonal Antibody
Antikörper
DCBLD2 antibody can be used in ELISA, and immunohistochemistry starting at 40 μg/mL.

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DCBLD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR