Anti-XRCC5 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-869EA 480 EUR
PRSI48-869
Anti-XRCC5 Mouse Monoclonal Antibody
Antikörper
Originally identified as an autoantigen, the Ku protein are nuclear proteins found in eukaryotes. They exist as a heterodimer consisiting of Ku 70 and Ku 80, migrating at approximately 70 and 80 kDa respectively. They bind to DNA ends at double strand breaks. The Ku proteins interact with the DNA-dependent protein kinase (DNA-PK) and participate in the targeting of DNA-PK to DNA. The Ku proteins may participate in numerous cellular processes, including DNA repair and replication, cell signalling, cell proliferation, transcriptional activation and apoptosis.

XRCC5 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

Protein G Column

PBS.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: XRCC5
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR