Anti-XAB2 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-865EA 480 EUR
PRSI48-865
Anti-XAB2 Rabbit Polyclonal Antibody
Antikörper
XAB2 (XPA-binding protein 2) was first identified through its interaction with XPA. Immunoprecipitation experiments have also demonstrated that a fraction of XAB2 interacts with the transcription-coupled repair-specific proteins CSA and CSB as well as with RNA polymerase II. Microinjection experiments with XAB2 antibodies resulted in the inhibition of both transcription-coupled repair and transcription.

XAB2 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 μg/mL, and immunoprecipitation

PBS.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: XAB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR