Anti-SAFB Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-862EA 480 EUR
PRSI48-862
Anti-SAFB Mouse Monoclonal Antibody
Antikörper
Hsp27 ERE-TATA binding protein (HET)/ Scaffold Attachment Factor-B (SAF-B) is a member of a subset of nuclear matrix proteins that mediate the attachment of chromatin to nuclear protein complexes. The calculated molecular weight of HET/SAF-B is 102 kDa (Accession No. U72355), but on SDS PAGE gels HET/SAF-B runs as an approximately 130 kDa protein. This aberrant migration is due to a large number of charged amino acid residues.

SAFB antibody can be used in Western Blot starting at 1:1000 - 1:3000, and immunohistochemistry starting at 10 μg/mL.

Protein G Column

PBS.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SAFB
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Gibbon, Gorilla, Human, Monkey, Orangutan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR